missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TEK Protein
A cDNA sequence encoding the TEK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13719-2ug
Additional Details : Weight : 0.01000kg
Specifications
TEK Protein | |
Research Use Only | |
Recombinant Protein | |
Mouse | |
MDSLAGLVLC GVSLLLYGVV EGAMDLILIN SLPLVSDAET SLTCIASGWH PHEPITIGRD FEALMNQHQD PLEVTQDVTR EWAKKVVWKR EKASKINGAY FCEGRVRGQA IRIRTMKMRQ QASFLPATLT MTVDRGDNVN ISFKKVLIKE EDAVIYKNGS FIHSVPRHEV PDILEVHLPH AQPQDAGVYS ARYIGGNLFT SAFTRLIVRR CEAQKWGPDC SRPCTTCKNN GVCHEDTGEC ICPPGFMGRT CEKACEPHTF GRTCKERCSG PEGCKSYVFC LPDPYGCSCA TGWRGLQCNE ACPSGYYGPD CKLRCHCTNE EICDRFQGCL CSQGWQGLQC EKEGRPRMTP QIEDLPDHIE VNSGKFNPIC KASGWPLPTS EEMTLVKPDG TVLQPNDFNY TDRFSVAIFT VNRVLPPDSG VWVCSVNTVA GMVEKPFNIS VKVLPEPLHA PNVIDTGHNF AIINISSEPY FGDGPIKSKK LFYKPVNQAW KYIEVTNEIF TLNYLEPRTD YELCVQLARP GEGGEGHPGP VRRFTTASIG LPPPRGLSLL PKSQTALNLT WQPIFTNSED EFYVEVERRS LQTTSDQQNI KVPGNLTSVL LSNLVPREQY TVRARVNTKA QGEWSEELRA WTLSDILPPQ PENIKISNIT DSTAMVSWTI VDGYSISSII IRYKVQGKNE DQHIDVKIKN ATVTQYQLKG LEPETTYHVD IFAENNIGSS NPAFSHELRT LPHSPASADL GGGKMLLIAI LGSAGMTCIT VLLAFLIMLQ LKRANVQRRM AQAFQNREEP AVQFNSGTLA LNRKAKNNPD PTIYPVLDWN DIKFQDVIGE GNFGQVLKAR IKKDGLRMDA AIKRMKEYAS KDDHRDFAGE LEVLCKLGHH PNIINLLGAC EHRGYLYLAI EYAPHGNLLD | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
TEK Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS. |