missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombospondin-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Thrombospondin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6785-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Thrombospondin-1 Protein | |
Research Use Only | |
Thbs1 | |
Recombinant Protein | |
E. coli | |
NRIPESGGDN GVFDIFELIG GARRGPGRRL VKGQDLSSPA FRIENANLIP AVPDDKFQDL LDAVWADKGF IFLASLRQMK KTRGTLLAVE RKDNTGQIFS VVSNGKAGTL DLSLSLPGKQ QVVSVEEALL ATGQWKSITL FVQEDRAQLY IDCDKMESAE LDVPIQSIFT RDLASVARLR VAKGDVNDNF QGVLQNVRFV FGTTPEDILR NKGCSSSTNV LLTLDNNVVN GSSPAIRTNY IGHKTKDLQA ICGLSCDELS SMVLELKGLR TIVTTLQDSI RKVTEENREL VSELKRPPLC FHNGVQYKNN EEWTVDSCTE CHCQNSVTIC KK | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |