missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Toll-like receptor 7 Protein

A DNA sequence encoding the Mus musculus (Mouse) Toll-like receptor 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6801-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001277684.1
Toll-like receptor 7 Protein
Research Use Only
Tlr7
Recombinant Protein
E. coli
FRWFPKTLPC EVKVNIPEAH VIVDCTDKHL TEIPEGIPTN TTNLTLTINH IPSISPDSFR RLNHLEEIDL RCNCVPVLLG SKANVCTKRL QIRPGSFSGL SDLKALYLDG NQLLEIPQDL PSSLHLLSLE ANNIFSITKE NLTELVNIET LYLGQNCYYR NPCNVSYSIE KDAFLVMRNL KVLSLKDNNV TAVPTTLPPN LLELYLYNNI IKKIQENDFN NLNELQVLDL SGNCPRCYNV PYPCTPCENN SPLQIHDNAF NSLTELKVLR LHSNSLQHVP PTWFKNMRNL QELDLSQNYL AREIEEAKFL HFLPNLVELD FS
Greater than 90% as determined by SDS-PAGE.
170743
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers