missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9425-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_064295.2 | |
Tryptophan 2,3-dioxygenase Protein | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56720 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
Yeast | |
MSGCPFAGNS VGYTLKNVSM EDNEEDRAQT GVNRASKGGL IYGNYLQLEK ILNAQELQSE VKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VIARMHRVVV IFKLLVQQFS VLETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQS LRVPYNRKHY RDNFGGDYNE LLLKSEQEQT LLQLVEAWLE RTPGLEPNGF NFWGKFEKNI LKGLEEEFLR IQAKTDSEEK EEQMAEFRKQ KEVLLCLFDE KRHDYLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDTL MTKWRYNHVC MVHRMLGTKA GTGGSSGYHY LRSTVSDRYK VFVDLFNLST YLVPRHWVPK MNPIIHKFLY TAEYSDSSYF SSDESD |