missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tyrosine-protein kinase JAK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6245-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Tyrosine-protein kinase JAK1 Protein | |
Research Use Only | |
JAK1 | |
Recombinant Protein | |
E. coli | |
EEQNPDIVSE KQPTTEVDPT HFEKRFLKRI RDLGEGHFGK VELCRYDPEG DNTGEQVAVK SLKPESGGNH IADLKKEIEI LRNLYHENIV KYKGICMEDG GNGIKLIMEF LPSGSLKEYL PKNKNKINLK QQLKYAIQIC KGMDYLGSRQ YVHRDLAARN VLVESEHQVK IGDFGLTKAI ETDKEYYTVK DDRDSPVFWY APECLIQCKF YIASDVWSFG VTLHELLTYC DSDFSPMALF LKMIGPTHGQ MTVTRLVKTL KEGKRLPCPP NCPDEVYQLM RKCWEFQPSN RTTFQNLIEG FEALL | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |