missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Ubiquitin-protein ligase E3A Protein

A DNA sequence encoding the Mus musculus (Mouse) Ubiquitin-protein ligase E3A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6865-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_035798.2
Ubiquitin-protein ligase E3A Protein
Research Use Only
Ube3a
Recombinant Protein
Yeast
HIIDDALVRL EMIAMENPAD LKKQLYVEFE GEQGVDEGGV SKEFFQLVVE EIFNPDIGMF TYDEATKLFW FNPSSFETEG QFTLIGILGL AIYNNCILDV HFPMVVYRKL MGKKGTFRDL GDSHPVLYQS LKDLLEYEGS VEDDMMITFQ ISQTDLFGNP MMYDLKENGD KIPITNENRK EFVISYSDYI LNKSVEKQFK AFRRGFHMVT NESPLKYLFR PEEIELLICG SRNLDFQALE ETTEYDGGYT RESVVIREFW EIVHSFTDEQ KRLFLLFTTG TDRAPVGGLG KLKMIIAKNG PDTERLPTSH TCFNVLLLPE YSSKEKLNV
Greater than 90% as determined by SDS-PAGE.
22215
200 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers