Learn More
enQuireBio™ Recombinant Mouse WNT1-inducible-signaling pathway protein 1
A DNA sequence encoding the Mus musculus (Mouse) WNT1-inducible-signaling pathway protein 2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8274-ye-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16093701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
NP_058569.2
WNT1-inducible-signaling pathway protein 1
Research Use Only
WISP2
Recombinant Protein
Yeast
QLCPAPCACP WTPPQCPPGV PLVLDGCGCC RVCARRLGES CDHLHVCDPS QGLVCQPGAG PSGRGAVCLF EEDDGSCEVN GRRYLDGETF KPNCRVLCRC DDGGFTCLPL CSEDVRLPSW DCPRPRRIQV PGRCCPEWVC DQAVMQPAIQ PSSAQGHQLS ALVTPASADG PCPNWSTAWG PCSTTCGLGI ATRVSNQNRF CQLEIQRRLC LSRPCLASRS HGSWNSAF
Greater than 90% as determined by SDS-PAGE.
22403
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol