Läs mer
enQuireBio™ Recombinant Mouse Zinc finger protein 346 Protein
Produktkod. 16026690
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16026690
Brand: enQuireBio™ QP6908ec200ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Mus musculus (Mouse) Zinc finger protein 346, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_036147.1
Gen-ID (Entrez) 26919
Namn Zinc finger protein 346 Protein
Kvantitet 200 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol Znf346
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Mouse
Art E. coli
Protein Tag His-SUMO
Sekvens MECPAPDATD AADPGEAGPY KGSEEPEGRE PDGVRFDRER ARRLWEAVSG AQPAGREEVE HMIQKNQCLF TSTQCKVCCA MLISESQKLA HYQSKKHANK VKRYLAIHGM ETIKGDVKRL DSDQKSSRSK DKNHCCPICN MTFSSPAVAQ SHYLGKTHAK SLKLKQQSTK GAALQQNREM LDPDKFCSLC HSTFNDPAMA QQHYMGKRHR KQETKLKLMA HYGRLADPAV SDLPAGKGYP CKTCKIVLNS IEQYQAHVSG FKHKNQSPKT LVTLGSQTPV QTQPTPKDSS TVQD
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.