Läs mer
enQuireBio™ Recombinant other Deslorelin Protein
Kvantitet:
5 mg
25 mg
100 mg
Förpackningsstorlek:
100mg
25mg
5mg
Specifikationer
Specifikationer
| Namn | Deslorelin Protein |
| Kvantitet | 100 mg |
| Regulatorisk status | Research Use Only |
| Endotoxinkoncentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Produkttyp | Recombinant Protein |
| Korsreaktivitet | Other |
| Art | E. coli |
| Protein Tag | Untagged |
| Buffert | The Deslorelin peptide was lyophilized with no additives. |
| Renhet eller kvalitetsklass | Greater than 99.0% as determined by RP-HPLC. |