missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PMSG Protein
A cDNA sequence encoding the PMSG was constructed and used to recombinantly synthesize the protein.
1295.00 SEK - 21610.00 SEK
Specifications
Name | PMSG Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942449
|
enQuireBio™
QP13081-100000IU |
100000 IU |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15952449
|
enQuireBio™
QP13081-1000IU |
1000 IU |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15962449
|
enQuireBio™
QP13081-5000IU |
5000 IU |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
PMSG Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The PMSG was lyophilized with no additives. |