missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Shiga Like Toxin 2 Protein
A cDNA sequence encoding the Shiga Like Toxin 2 was constructed and used to recombinantly synthesize the protein.
2690.00 SEK - 20560.00 SEK
Specifications
Name | Shiga Like Toxin 2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913759
|
enQuireBio™
QP13496-100UG |
100 μg |
2690.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933759
|
enQuireBio™
QP13496-500UG |
500 μg |
10265.00 SEK
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923759
|
enQuireBio™
QP13496-1MG |
1 mg |
20560.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Shiga Like Toxin 2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Shiga Like Toxin 2 protein solution contains PBS, 50mM arginine and 0.05% sodium azide. |