missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. gingivalis Gingipain R1 Protein

A DNA sequence encoding the Porphyromonas gingivalis Gingipain R1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7192-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087640

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

P. gingivalis Gingipain R1 Protein
Research Use Only
rgpA
Recombinant Protein
Yeast
YTPVEEKQNG RMIVIVAKKY EGDIKDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENVI ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLVNYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVACVNGDFL FSMPCFAEAL MRAQKDGKPT GTVAIIASTI NQSWASPMRG QDEMNEILCE KHPNNIKRTF GGVTMNGMFA MVEKYKKDGE KMLDTWTVFG DPSLLVRTLV PTKMQVTAPA QINLTDASVN VSCDYNGAIA TISANGKMFG SAVVENGTAT INLTGLTNES TLTLTVVGYN KETVIKTINT NGEPNPYQPV SNLTATTQGQ KVTLKWDAPS TKTNATTNTA RSVDGIRELV LLSVSDAPEL LRS
Greater than 80% as determined by SDS-PAGE.
200 μg
Not determined.
Partial (see sequence information for more details).
P. gingivalis
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers