missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. gingivalis Lys-gingipain Protein
A DNA sequence encoding the Porphyromonas gingivalis (strain ATCC 33277 / DSM 20709 / JCM 12257) Lys-gingipain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7438-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_012458488.1 | |
P. gingivalis Lys-gingipain Protein | |
Research Use Only | |
kgp | |
Recombinant Protein | |
E. coli | |
DVYTDHGDLY NTPVRMLVVA GAKFKEALKP WLTWKAQKGF YLDVHYTDEA EVGTTNASIK AFIHKKYNDG LAASAAPVFL ALVGDTDVIS GEKGKKTKKV TDLYYSAVDG DYFPEMYTFR MSASSPEELT NIIDKVLMYE KATMPDKSYL EKALLIAGAD SYWNPKIGQQ TIKYAVQYYY NQDHGYTDVY SYPKAPYTGC YSHLNTGVGF ANYTAHGSET SWADPSLTAT QVKALTNKDK YFLAIGNCCV TAQFDYPQPC FGEVMTRVKE KGAYAYIGSS PNSYWGEDYY WSVGANAVFG VQPTFEGTSM GSYDATFLED SYNTVNSIMW AGNLAATHAG NIGNITHIGA HYYWEAYHVL GDGSVM | |
Greater than 82.5% as determined by SDS-PAGE. |
29256891 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
P. gingivalis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |