missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
enQuireBio™ Recombinant P. gingivalis Peptidylarginine deiminase Protein
Specifications
Specifications
| Tillträdesnummer | WP_005873463.1 |
| Gen-ID (Entrez) | 29256112 |
| Namn | P. gingivalis Peptidylarginine deiminase Protein |
| Kvantitet | 100 μg |
| Regulatorisk status | Research Use Only |
| Endotoxinkoncentration | Not determined. |
| Gensymbol | PG_1424 |
| Protein Längd | Full Length |
| Produkttyp | Recombinant Protein |
| Korsreaktivitet | P. gingivalis |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?