Learn More
enQuireBio™ Recombinant Pseudomonas Creatinine amidohydrolase Protein
A DNA sequence encoding the Pseudomonas putida (Arthrobacter siderocapsulatus) Creatinine amidohydrolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6914-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Pseudomonas Creatinine amidohydrolase Protein | |
Research Use Only | |
crnA | |
Recombinant Protein | |
E. coli | |
MSKSVFVGEL TWKEYEARVA AGDCVLMLPV GALEQHGHHM CMNVDVLLPT AVCKRVAERI GALVMPGLQY GYKSQQKSGG GNHFPGTTSL DGATLTGTVQ DIIRELARHG ARRLVLMNGH YENSMFIVEG IDLALRELRY AGIQDFKVVV LSYWDFVKDP AVIQQLYPEG FLGWDIEHGG VFETSLMLAL YPDLVDLDRV VDHPPATFPP YDVFPVDPAR TPAPGTLSSA KTASREKGEL ILEVCVQGIA DAIREEFPPT | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
seudomonas | |
GST | |
Tris-based buffer, 50% glycerol |