missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat BDNF Protein Isoform 2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) BDNF Protein Isoform 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8869-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056911

  • 6002.00 SEK / 100µg


Estimated Shipment
09-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001257559.1
BDNF Protein Isoform 2 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
24225
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BDNF
Recombinant Protein
E. coli
RRGELSVCDS ISEWVTAADK KTAVDMSGGT VTVLEKVPVS KGQLKQYFYE TKCNPMGYTK EGCRGIDKRH WNSQCRTTQS YVRALTMDSK KRIGWRFIRI DTSCVCTL
Videos
SDS
Documents