missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL13 / BCA-1 / BLC Protein
A DNA sequence encoding the Rattus norvegicus (Rat) CXCL13 / BCA-1 / BLC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8592-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001017496.1 | |
CXCL13 / BCA-1 / BLC Protein | |
100 μg | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |
498335 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CXCL13 | |
Recombinant Protein | |
E. coli | |
LETHYTNLKC RCSKVSSTFI NLILVDWIQV IRPGNGCPKT EIIFWTKAKK AICVNPTARW LPKVLKFVRR SITSTPQAPV SKKRAA |