missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Rat E-Cadherin Fc Chimera Protein

Produktkod. 18798867
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18798867 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18798867 Leverantör R&D Systems™ Leverantörsnummer 8144EC050
 Begränsad artikel
Beräknad leverans: 03-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Rat E-Cadherin Fc Chimera Protein is derived from NS0. The Recombinant Rat E-Cadherin Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifikationer

Formulering Lyophilized from a 0.2μm filtered solution in PBS.
Gen-ID (Entrez) 83502
Molekylvikt (g/mol) 103 kDa (pro monomer) & 87 kDa (mature monomer)
Namn E-Cadherin Fc Chimera
Kvantitet 50 μg
Källa Mouse myeloma cell line,NS0-derived rat E-Cadherin protein, (N-terminus) Rat E-Cadherin (Gln 24-Ser704) Accession &Num; Q9R0T4 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Förvaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturell form Disulfide-linked homodimer
Konjugera Unconjugated
Rekombinant Recombinant
Produktlinje R&D Systems
Renhet eller kvalitetsklass 95%, by SDS-PAGE with silver staining.
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.