missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FABP3 / H-FABP Protein
A DNA sequence encoding the Rattus norvegicus (Rat) FABP3 / H-FABP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6013-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_077076.1 | |
FABP3 / H-FABP Protein | |
100 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79131 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Fabp3 | |
Recombinant Protein | |
E. coli | |
ADAFVGTWKL VDSKNFDDYM KSLGVGFATR QVASMTKPTT IIEKNGDTIT IKTHSTFKNT EISFQLGVEF DEVTADDRKV KSVVTLDGGK LVHVQKWDGQ ETTLTRELSD GKLILTLTHG NVVSTRTYEK EA |