missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Galectin-4 Protein

Product Code. 16002270
Click to view available options
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16002270

Brand: enQuireBio™ QP6297ec1mg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) Galectin-4, was expressed in the hosts and tags indicated.

Specifications

Tillträdesnummer NP_037107.1
Gen-ID (Entrez) 25474
Namn Galectin-4 Protein
Kvantitet 1 mg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol Lgals4
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Rat
Art E. coli
Protein Tag His-SUMO
Sekvens MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSIYIQGIAK DNMRRFHVNF AVGQDEGADI AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGHH FELVFMVMSE HYKVVVNGTP FYEYGHRLPL QMVTHLQVDG DLELQSINFL GGQPAASQYP GTMTIPAYPS AGYNPPQMNS LPVMAGPPIF NPPVPYVGTL QGGLTARRTI IIKGYVLPTA KNLIINFKVG STGDIAFHMN PRIGDCVVRN SYMNGSWGSE ERKIPYNPFG AGQFFDLSIR CGTDRFKVFA NGQHLFDFSH RFQAFQRVDM LEIKGDITLS YVQI
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.