missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Haptoglobin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Haptoglobin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8887-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_036714.2 | |
Haptoglobin Protein | |
Research Use Only | |
HP | |
Recombinant Protein | |
E. coli | |
VELGNDATDI EDDSCPKPPE IANGYVEHLV RYRCRQFYKL QTEGDGIYTL NSEKQWVNPA AGDKLPKCEA VCGKPKHPVD QVQRIIGGSM DAKGSFPWQA KMISRHGLTT GATLISDQWL LTTAQNLFLN HSENATAKDI APTLTLYVGK NQLVEIEKVV LHPERSVVDI GLIKLKQKVL VTEKVMPICL PSKDYVAPGR MGYVSGWGRN VNFRFTERLK YVMLPVADQE KCELHYEKST VPEKKGAVSP VGVQPILNKH TFCAGLTKYE EDTCYGDAGS AFAVHDTEED TWYAAGILSF DKSCAVAEYG VYVRATDLKD WVQETMAK | |
Greater than 90% as determined by SDS-PAGE. |
24464 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |