missing translation for 'onlineSavingsMsg'
Läs mer

enQuireBio™ Recombinant Rat Hemopexin / HPX Protein

Produktkod. 16081370
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16081370

Brand: enQuireBio™ QP6180ye10ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) Hemopexin / HPX, was expressed in the hosts and tags indicated.

Specifications

Tillträdesnummer NP_445770.1
Gen-ID (Entrez) 58917
Namn Hemopexin / HPX Protein
Kvantitet 10 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol HPX
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Rat
Art Yeast
Protein Tag His
Sekvens NPLPAAHETV AKGENGTKPD SDVIEHCSDA WSFDATTMDH NGTMLFFKGE FVWRGHSGIR ELISERWKNP VTSVDAAFRG PDSVFLIKED KVWVYPPEKK ENGYPKLFQE EFPGIPYPPD AAVECHRGEC QSEGVLFFQG NRKWFWDFAT RTQKERSWPA VGNCTAALRW LERYYCFQGN KFLRFNPVTG EVPPRYPLDA RDYFISCPGR GHGKLRNGTA HGNSTHPMHS RCNADPGLSA LLSDHRGATY AFSGSHYWRL DSSRDGWHSW PIAHHWPQGP SAVDAAFSWD EKVYLIQGTQ VYVFLTKGGN NLVSGYPKRL EKELGSPPGI SLDTIDAAFS CPGSSKLYVT SGRRLWWLDL KSGAQATWAE LSWPHEKVDG ALCLEKSLGP YSCSSNGPNL FFIHGPNLYC YSSIDKLNAA KSLPQPQKVN SILGCSQ
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.