missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL-18 / IL-1F4 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) IL-18 / IL-1F4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8901-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_062038.1 | |
IL-18 / IL-1F4 Protein | |
Research Use Only | |
IL18 | |
Recombinant Protein | |
E. coli | |
HFGRLHCTTA VIRSINDQVL FVDKRNPPVF EDMPDIDRTA NESQTRLIIY MYKDSEVRGL AVTLSVKDGR MSTLSCKNKI ISFEEMNPPE NIDDIKSDLI FFQKRVPGHN KMEFESSLYE GHFLACQKED DAFKLVLKRK DENGDKSVMF TLTNLHQS | |
Greater than 90% as determined by SDS-PAGE. |
29197 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |