missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 9 Protein
A cDNA sequence encoding the IL 9 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 80310.00 SEK
Specifications
Gene ID (Entrez) | 116558 |
---|---|
Name | IL 9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944968
|
enQuireBio™
QP10721-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924968
|
enQuireBio™
QP10721-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934968
|
enQuireBio™
QP10721-1MG |
1 mg |
80310.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
116558 | |
Research Use Only | |
Il9 | |
Recombinant Protein | |
E. coli | |
MQRCSTSWGI QHTSYLIENL KDDPSSKCSC SANVTSCLCL PIPSDDCTTP CFQEGMSQVT NATQQSKFSP FFFRVKRIVE TLKSNKCQFF SCEKPCNQTT AGNTVSFLKS LLKTFQKTEV QVQRSRA | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 9 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine TS1 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Rat | |
Untagged | |
IL9 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |