missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Kinesin-like protein KIF1C Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Kinesin-like protein KIF1C, was expressed in the hosts and tags indicated.
2430.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_665884.1 |
---|---|
Gene ID (Entrez) | 113886 |
Name | Kinesin-like protein KIF1C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16052020
|
enQuireBio™
QP6263-YE-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092020
|
enQuireBio™
QP6263-YE-500UG |
500 μg |
24845.00 SEK
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062020
|
enQuireBio™
QP6263-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082020
|
enQuireBio™
QP6263-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002030
|
enQuireBio™
QP6263-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072020
|
enQuireBio™
QP6263-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_665884.1 | |
Kinesin-like protein KIF1C Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
113886 | |
Research Use Only | |
Kif1c | |
Recombinant Protein | |
Yeast | |
MAGASVKVAV RVRPFNARET SQDAKCVVSM QGNTTSIINP KQSRMFLKAS FDYSYWSHTS VEDPQFASQQ QVYRDIGEEM LLHAFEGYNV CIFAYGQTGA GKSYTMMGRQ EPGQQGIVPQ LCEDLFSRVN VNQSAQLSYS VEVSYMEIYC ERVRDLLNPK SRGSLRVREH PILGPYVQDL SKLAVTSYAD IADLMDCGNK ARTVAATNMN ETSSRSHAVF TIVFTQRSHD QLTGLDSEKV SKISLVNLAG SERADSSGAR GMRLKEGANI NK | |
Greater than 90% as determined by SDS-PAGE. |