missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat LDHA Protein
A cDNA sequence encoding the LDHA was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 20560.00 SEK
Specifications
Gene ID (Entrez) | 24533 |
---|---|
Name | LDHA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Ldha |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980779
|
enQuireBio™
QP12549-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970779
|
enQuireBio™
QP12549-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960779
|
enQuireBio™
QP12549-100UG |
100 μg |
20560.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
24533 | |
Research Use Only | |
Ldha | |
Rat | |
His | |
LDHA protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 1mM DTT and 20% glycerol. |
LDHA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MAALKDQLIV NLLKEEQVPQ NKITVVGVGA VGMACAISIL MKDLADELAL VDVIEDKLKG EMMDLQHGSL FLKTPKIVSS KDYSVTANSK LVIITAGARQ QEGESRLNLV QRNVNIFKFI IPNVVKYSPQ CKLLIVSNPV DILTYVAWKI SGFPKNRVIG SGCNLDSARF RYLMGERLGV HPLSCHGWVL GEHGDSSVPV WSGVNVAGVS LKSLNPQLGT DADKEQWKDV HKQVVDSAYE VIKLKGYTSW AIGLSVADLA ESIMKNLRRV HPISTMIKGL YGIKEDVFLS VPCILGQNGI SDVVKVTLTP DEEARLKKSA DTLWGIQKEL QFLEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |