missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Monoglyceride lipase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Monoglyceride lipase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9272-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_612511.1 | |
Monoglyceride lipase Protein | |
Research Use Only | |
Mgll | |
Recombinant Protein | |
Yeast | |
MPEASSPRRT PQNVPYQDLP HLVNADGQYL FCRYWKPSGT PKALIFVSHG AGEHCGRYDE LAQMLKRLDM LVFAHDHVGH GQSEGERMVV SDFQVFVRDL LQHVNTVQKD YPEVPVFLLG HSMGGAISIL AAAERPTHFS GMILISPLIL ANPESASTLK VLAAKLLNFV LPNISLGRID SSVLSRNKSE VDLYNSDPLI CHAGVKVCFG IQLLNAVSRV ERAMPRLTLP FLLLQGSADR LCDSKGAYLL MESSPSQDKT LKMYEGAYHV LHKELPEVTN SVLHEINTWV SHRIAVAGAR CLP | |
Greater than 90% as determined by SDS-PAGE. |
29254 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |