missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PCSK1 / NEC1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PCSK1 / NEC1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9322-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_058787.1
PCSK1 / NEC1 Protein
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
25204
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Pcsk1
Recombinant Protein
Yeast
SVPRDSALNL FNDPMWNQQW YLQDTRMTAS LPKLDLHVIP VWQKGITGKG VVITVLDDGL EWNHTDIYAN YDPEASYDFN DNDHDPFPRY DPTNENKHGT RCAGEIAMQA NNHKCGVGVA YNSKVGGIRM LDGIVTDAIE ASSIGFNPGH VDIYSASWGP NDDGKTVEGP GRLAQKAFEY GVKQGRQGKG SIFVWASGNG GRQGDNCDCD GYTDSIYTIS ISSASQQGLS PWYAEKCSST LATSYSSGDY TDQRITSADL HNDCTETHTG TSASAPLAAG IFALALEANP NLTWRDMQHL VVWTSEYDPL ANNPGWKKNG AGLMVNSRFG FGLLNAKALV DLADPRTWRN VPEKKECIIK DNNFEPRALK ANGEVIVEIP TRACEGQENA INSLEHVQFE ATIEYSRRGD LHVTLTSAAG TSTVLLAERE RDTSPNGFKN WDFMSVHTWG ENPVGTWTLK VTDMSGRMQN EGRIVNWKLI LHGTSSQPEH MKQPRVYTSY NTVQNDRRGV EKMVNVVEEK PTQNSLNGNL LVPKNSSSSS VEDRRDEQVQ GAPSKAMLRL LQSAFSKNTP SKQSSKIPSA KLSVPYEGLY EALEKLNKPS QLEDSEDSLY SDYVDVFYNT KPYKHRDDRL LQALMDILNE KN
Videos
SDS
Documents