missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Prolactin-inducible protein homolog Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Prolactin-inducible protein homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9736-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_073199.1 | |
Prolactin-inducible protein homolog Protein | |
Research Use Only | |
Pip | |
Recombinant Protein | |
Yeast | |
QDNETIPQPL LFQLNVPSTP DENQEVDMSL TLQTQYKECL VVKAYLISNT PVDGGFNYIQ TRCICNDHPT TLYWTFVVTQ TLTFRIMVDI VKDKGICPNN VAVVPISGNR YFTDRTVYVN | |
Greater than 90% as determined by SDS-PAGE. |
64673 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |