missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Protein-lysine 6-oxidase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Protein-lysine 6-oxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6314-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092400

  • 8695.00 SEK / 100µg
Estimated Shipment: 22-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_058757.1
Protein-lysine 6-oxidase Protein
Research Use Only
Lox
Recombinant Protein
E. coli
DDPYNPYKYS DDNPYYNYYD TYERPRSGSR HRPGYGTGYF QYGLPDLVPD PYYIQASTYV QKMSMYNLRC AAEENCLASS AYRADVRDYD HRVLLRFPQR VKNQGTSDFL PSRPRYSWEW HSCHQHYHSM DEFSHYDLLD ASTQRRVAEG HKASFCLEDT SCDYGYHRRF ACTAHTQGLS PGCYDTYAAD IDCQWIDITD VQPGNYILKV SVNPSYLVPE SDYSNNVVRC EIRYTGHHAY ASGCTISPY
Greater than 90% as determined by SDS-PAGE.
24914
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers