missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Regucalcin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Regucalcin, was expressed in the hosts and tags indicated.
2515.00 SEK - 26260.00 SEK
Specifications
Accession Number | NP_113734.1 |
---|---|
Gene ID (Entrez) | 25106 |
Name | Regucalcin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034470
|
enQuireBio™
QP6601-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074470
|
enQuireBio™
QP6601-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024470
|
enQuireBio™
QP6601-YE-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054470
|
enQuireBio™
QP6601-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064470
|
enQuireBio™
QP6601-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044470
|
enQuireBio™
QP6601-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_113734.1 | |
Regucalcin Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
25106 | |
Research Use Only | |
Rgn | |
Recombinant Protein | |
Yeast | |
MSSIKIECVL RENYRCGESP VWEEASKCLL FVDIPSKTVC RWDSISNRVQ RVGVDAPVSS VALRQSGGYV ATIGTKFCAL NWEDQSVFIL AMVDEDKKNN RFNDGKVDPA GRYFAGTMAE ETAPAVLERH QGSLYSLFPD HSVKKYFDQV DISNGLDWSL DHKIFYYIDS LSYTVDAFDY DLPTGQISNR RTVYKMEKDE QIPDGMCIDV EGKLWVACYN GGRVIRLDPE TGKRLQTVKL PVDKTTSCCF GGKDYSEMYV TCARDGMSAE GLLRQPDAGN IFKITGLGVK GIAPYSYAG | |
Greater than 90% as determined by SDS-PAGE. |