missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat S100A8 / CAGA Protein

A DNA sequence encoding the Rattus norvegicus (Rat) S100A8 / CAGA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6642-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064750

  • 26542.00 SEK / 1mg


Estimated Shipment
09-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_446274.2
S100A8 / CAGA Protein
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
116547
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S100A8
Recombinant Protein
E. coli
ATELEKALSN VIEVYHNYSG IKGNHHALYR DDFRKMVTTE CPQFVQNKNT ESLFKELDVN SDNAINFEEF LVLVIRVGVA AHKDSHKE
Videos
SDS
Documents