missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinA1 / A1AT Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinA1 / A1AT, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_071964.2 |
---|---|
Gene ID (Entrez) | 24648 |
Name | SerpinA1 / A1AT Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044930
|
enQuireBio™
QP6666-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064930
|
enQuireBio™
QP6666-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054930
|
enQuireBio™
QP6666-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074930
|
enQuireBio™
QP6666-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084930
|
enQuireBio™
QP6666-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094930
|
enQuireBio™
QP6666-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_071964.2 | |
SerpinA1 / A1AT Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24648 | |
Research Use Only | |
Serpina1 | |
Recombinant Protein | |
E. coli | |
EDAQETDTSQ QDQSPTYRKI SSNLADFAFS LYRELVHQSN TSNIFFSPMS ITTAFAMLSL GSKGDTRKQI LEGLEFNLTQ IPEADIHKAF HHLLQTLNRP DSELQLNTGN GLFVNKNLKL VEKFLEEVKN NYHSEAFSVN FADSEEAKKV INDYVEKGTQ GKIVDLMKQL DEDTVFALVN YIFFKGKWKR PFNPEHTRDA DFHVDKSTTV KVPMMNRLGM FDMHYCSTLS SWVLMMDYLG NATAIFLLPD DGKMQHLEQT LTKDLISRFL LNRQTRSAIL YFPKLSISGT YNLKTLLSSL GITRVFNNDA DLSGITEDAP LKLSQAVHKA VLTLDERGTE AAGATVVEAV PMSLPPQVKF DHPFIFMIVE SETQSPLFVG KVIDPTR | |
Greater than 90% as determined by SDS-PAGE. |