missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat SerpinB2 / PAI-2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) SerpinB2 / PAI-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6669-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074960

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_067728.1
SerpinB2 / PAI-2 Protein
Research Use Only
Serpinb2
Recombinant Protein
E. coli
MEELSMANTM FALNLLKQIE QSNSTQNIFI SPWSISSTLA IVFLGAQANT EEQMAKVLNF DKIGSYDLTP GNPENFHGCD FAQHIQRDNY PVAILQAQAR DKIHSAFSSL SSTINTPRLG DYLLESANKL FGEKSARFKE EYIQRCKKYY STEPEAVDFL ECANEARKKI NSWVKTQTKG EIPNLLPEGS VDEDTKMVLV NTIYFKGRWK TPFQKRLNGL YPFRVNLNES KPVQMMYLRE KLNIGYIKDL KTQILELPYI GNISMFLLLP DEIEDSSTGL EMLEREINFD NFNKWISKET LDEDDVLVYI PKFKLAQNYE LKPILQRMGM EDAFNKGKAD FSGMSESNDL FLSEVFHQAT VDVNEEGTVA AGGTGAVMTG RTGHGGPQFV ADHPFLFFIM NNITRTILFV GRFSSP
Greater than 90% as determined by SDS-PAGE.
60325
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers