missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinE1 / PAI-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinE1 / PAI-1, was expressed in the hosts and tags indicated.
2515.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_036752.1 |
---|---|
Gene ID (Entrez) | 24617 |
Name | SerpinE1 / PAI-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16005000
|
enQuireBio™
QP6673-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025000
|
enQuireBio™
QP6673-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16015000
|
enQuireBio™
QP6673-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035000
|
enQuireBio™
QP6673-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045000
|
enQuireBio™
QP6673-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055000
|
enQuireBio™
QP6673-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036752.1 | |
SerpinE1 / PAI-1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24617 | |
Research Use Only | |
Serpine1 | |
Recombinant Protein | |
E. coli | |
SPLPESHTAQ QATNFGVKVF QHVVQASKDR NVVFSPYGVS SVLAMLQLTT AGKTRQQIQD AMGFNISERG TAPALRKLSK ELMGSWNKNE ISTADAIFVQ RDLELVQGFM PHFFKLFRTT VKQVDFSEVE RARFIINDWV ERHTKGMISD LLAKGAVNEL TRLVLVNALY FNGQWKTPFL EASTHQRLFH KSDGSTISVP MMAQNNKFNY TEFTTPDGHE YDILELPYHG ETLSMFIAAP FEKDVPLSAI TNILDAELIR QWKSNMTRLP RLLILPKFSL ETEVDLRGPL EKLGMTDIFS STQADFTSLS DQEQLSVAQA LQKVKIEVNE SGTVASSSTA ILVSARMAPT EMVLDRSFLF VVRHNPTETI LFMGQLMEP | |
Greater than 90% as determined by SDS-PAGE. |