missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinF1 / PEDF Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinF1 / PEDF, was expressed in the hosts and tags indicated.
2515.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_808788.1 |
---|---|
Gene ID (Entrez) | 287526 |
Name | SerpinF1 / PEDF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025010
|
enQuireBio™
QP6675-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045010
|
enQuireBio™
QP6675-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035010
|
enQuireBio™
QP6675-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055010
|
enQuireBio™
QP6675-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065010
|
enQuireBio™
QP6675-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075010
|
enQuireBio™
QP6675-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_808788.1 | |
SerpinF1 / PEDF Protein | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |
287526 | |
Research Use Only | |
Serpinf1 | |
Recombinant Protein | |
E. coli | |
NVPDSSQDSP APDSTGEPVV EEDDPFFKAP VNKLAAAVSN FGYDLYRLRS GAVSTGNILL SPLSVATALS ALSLGAEQRT ESVIHRALYY DLINNPDIHS TYKELLASVT APEKNFKSAS RIVFERKLRV KSSFVAPLEK SYGTRPRILT GNPRIDLQEI NNWVQAQMKG KIARSTREMP SALSILLLGV AYFKGQWATK FDSRKTTLQD FHLDEDRTVR VPMMSDPKAI LRYGLDSDLN CKIAQLPLTG SMSIIFFLPL TVTQNLTMIE ESLTSEFVHD IDRELKTIQA VLTVPKLKLS YEGDVTNSLQ DMKLQSLFES PDFSKITGKP VKLTQVEHRA AFEWNEEGAG TSSNPDLQPV RLTFPLDYHL NRPFIFVLRD TDTGALLFIG RILDPSST | |
Greater than 90% as determined by SDS-PAGE. |