missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Thrombopoietin / THPO Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Thrombopoietin / THPO, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8583-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015411

  • 8695.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_112395.1
Thrombopoietin / THPO Protein
Research Use Only
THPO
Recombinant Protein
E. coli
SPVPPACDPR LLNKLLRDSY LLHSRLSQCP DVNPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSALFLS LQQLLRGKVR FLLLVEGPAL CVRRTLPTTA VPSRTSQLLT LNK
Greater than 90% as determined by SDS-PAGE.
108348162
100 μg
Not determined.
Partial (see sequence information for more details).
Rat
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers