missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 16 Protein
A cDNA sequence encoding the IL 16 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10694-2ug
Additional Details : Weight : 0.01000kg
Specifications
IL 16 Protein | |
Research Use Only | |
Fully biologically active when compared to standard. Determined by its ability to chemoattract human CD4+ T-Lymphocytes using a concentration range of 50.0-100.0ng/ml corresponding to a Specific Activity of 10,000-20,000IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2?m filtered concentrated solution in PBS, pH 7.4. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SAASASAASD VSVESSAEAT VYTVTLEKMS AGLGFSLEGG KGSLHGDKPL TINRIFKGAA SEQSETIQPG DEILQLAGTA MQGLTRFEAW NIIKALPDGP VTIVIRRKSL QPKETTAAAD S | |
Greater than 98.0% as determined by SDS-PAGE. |