missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rotavirus Intermediate capsid protein VP6 Protein
A DNA sequence encoding the Rotavirus A (strain Cow/United Kingdom/UK/1975 G6-P7[5]-I2-R2-C2-M2-A3-N2-T7-E2-H3) (RV-A) Intermediate capsid protein VP6, was expressed in the hosts and tags indicated.
3615.00 SEK - 32610.00 SEK
Specifications
Name | Rotavirus Intermediate capsid protein VP6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | VP6 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058441
|
enQuireBio™
QP9540-YE-100UG |
100 μg |
12410.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098441
|
enQuireBio™
QP9540-YE-500UG |
500 μg |
29995.00 SEK
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068441
|
enQuireBio™
QP9540-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088441
|
enQuireBio™
QP9540-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008451
|
enQuireBio™
QP9540-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078441
|
enQuireBio™
QP9540-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Rotavirus Intermediate capsid protein VP6 Protein | |
Not determined. | |
Full Length | |
Rotavirus | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
VP6 | |
Recombinant Protein | |
Yeast | |
MDVLYSLSKT LKDARDKIVE GTLYSNVSDL IQQFNQMIIT MNGNEFQTGG IGNLPIRNWN FDFGLLGTTL LNLDANYVET ARNTIDYFVD FVDNVCMDEM VRESQRNGIA PQSDSLRKLS GIKFKRINFD NSSEYIENWN LQNRRQRTGF TFHKPNIFPY SASFTLNRSQ PAHDNLMGTM WLNAGSEIQV AGFDYSCAIN APANTQQFEH IVQLRRVLTT ATITLLPDAE RFSFPRVINS ADGATTWYFN PVILRPNNVE VEFLLNGQII NTYQARFGTI IARNFDTIRL SFQLMRPPNM TPAVAALFPN AQPFEHQATV GLTLRIESAV CESVLADASE TMLANVTSVR QEYAIPVGPV FPPGMNWTDL ITNYSPSRED NLQRVFTVAS IRSMLVK | |
Greater than 90% as determined by SDS-PAGE. |