Learn More
enQuireBio™ Recombinant Rotavirus Intermediate capsid protein VP6 Protein
A DNA sequence encoding the Rotavirus A (strain Cow/United Kingdom/UK/1975 G6-P7[5]-I2-R2-C2-M2-A3-N2-T7-E2-H3) (RV-A) Intermediate capsid protein VP6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9540-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
Rotavirus Intermediate capsid protein VP6 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Rotavirus | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
VP6 | |
Recombinant Protein | |
Yeast | |
MDVLYSLSKT LKDARDKIVE GTLYSNVSDL IQQFNQMIIT MNGNEFQTGG IGNLPIRNWN FDFGLLGTTL LNLDANYVET ARNTIDYFVD FVDNVCMDEM VRESQRNGIA PQSDSLRKLS GIKFKRINFD NSSEYIENWN LQNRRQRTGF TFHKPNIFPY SASFTLNRSQ PAHDNLMGTM WLNAGSEIQV AGFDYSCAIN APANTQQFEH IVQLRRVLTT ATITLLPDAE RFSFPRVINS ADGATTWYFN PVILRPNNVE VEFLLNGQII NTYQARFGTI IARNFDTIRL SFQLMRPPNM TPAVAALFPN AQPFEHQATV GLTLRIESAV CESVLADASE TMLANVTSVR QEYAIPVGPV FPPGMNWTDL ITNYSPSRED NLQRVFTVAS IRSMLVK |