missing translation for 'onlineSavingsMsg'
Läs mer

enQuireBio™ Recombinant S. aureus clpC Protein

Produktkod. 16032141
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16032141

missing translation for 'mfr': enQuireBio™ QP8010ec50ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) clpC, was expressed in the hosts and tags indicated.

Specifications

Tillträdesnummer WP_000897132.1
Gen-ID (Entrez) 31213331
Namn S. aureus clpC Protein
Kvantitet 50 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol clpC
Protein Längd Partial (see sequence information for more details).
Produkttyp Recombinant Protein
Korsreaktivitet S. aureus
Art E. coli
Protein Tag His-SUMO
Sekvens MLFGRLTERA QRVLAHAQEE AIRLNHSNIG TEHLLLGLMK EPEGIAAKVL ESFNITEDKV IEEVEKLIGH GQDHVGTLHY TPRAKKVIEL SMDEARKLHH NFVGTEHILL GLIRENEGVA ARVFANLDLN ITKARAQVVK ALGNPEM
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.