missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Clumping factor B Protein

A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) Clumping factor B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7759-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060631

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000745917.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
clfB
Recombinant Protein
E. coli
SEQSNDTTQS SKNNASADSE KNNTIETPQL NTTANDTSDI SANTNSANVD STAKTMSTQT SNTTTTEPAS TNETPQPTAI KDQATAAKMQ DQTVPQEANS QVDNKTTNDA NNIATNSELK NPQTLDLPQS SPQTISNAQG TSKPSVRTRA VRSLAVAEPV VNAADAKGTN VNDKVTASDF KLEKTAFDPN QSGNTFMAAN FKVTGQVKSG DYFTAKLPDS VTGNGDVDYS NSNNTMPIAD IKSTNGDVVA KATYDILTKT YTFVFTDYVN DKENINGQFS LPLFTDRAKA PKSGTYDANI NIADEMFDNK ITYNYSSPIA GIDKPNGANI SSQIIGVDTA SGQNTYKQTV FVNPKQRVLG NTWVYIKGYQ DKIEESSGKV SATDTKLRIF EVNDTSKLSD SYYADPNDSN LKEVTGEFKD KISYKYDNVA SINFGDINKT YVVLVEGHYD NTGKNLKTQV IQENIDPATG KDYSIFGWNN ENVVRYGGGS ADGDSAVN
S. aureus Clumping factor B Protein
10 μg
Not determined.
Partial (see sequence information for more details).
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents