missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type C-1 Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type C-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7401-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001043549.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
entC1 | |
Recombinant Protein | |
E. coli | |
ESQPDPTPDE LHKASKFTGL MENMKVLYDD HYVSATKVKS VDKFLAHDLI YNISDKKLKN YDKVKTELLN EGLAKKYKDE VVDVYGSNYY VNCYFSSKDN VGKVTGGKTC MYGGITKHEG NHFDNGNLQN VLIRVYENKR NTISFEVQTD KKSVTAQELD IKARNFLINK KNLYEFNSSP YETGYIKFIE NNGNTFWYDM MPAPGDKFDQ SKYLMMYNDN KTVDSKSVKI EVHLTTKNG |
S. aureus Enterotoxin type C-1 Protein | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |