missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type E Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7046-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
S. aureus Enterotoxin type E Protein | |
Research Use Only | |
entE | |
Recombinant Protein | |
E. coli | |
SEEINEKDLR KKSELQRNAL SNLRQIYYYN EKAITENKES DDQFLENTLL FKGFFTGHPW YNDLLVDLGS KDATNKYKGK KVDLYGAYYG YQCAGGTPNK TACMYGGVTL HDNNRLTEEK KVPINLWIDG KQTTVPIDKV KTSKKEVTVQ ELDLQARHYL HGKFGLYNSD SFGGKVQRGL IVFHSSEGST VSYDLFDAQG QYPDTLLRIY RDNKTINSEN LHIDLYLYTT | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |