missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus LDH-A / LDHA Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) LDH-A / LDHA, was expressed in the hosts and tags indicated.
3180.00 SEK - 38570.00 SEK
Specifications
Accession Number | WP_001031880.1 |
---|---|
Gene ID (Entrez) | 31213039 |
Name | S. aureus LDH-A / LDHA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057910
|
enQuireBio™
QP7260-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077910
|
enQuireBio™
QP7260-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067910
|
enQuireBio™
QP7260-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087910
|
enQuireBio™
QP7260-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097910
|
enQuireBio™
QP7260-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007920
|
enQuireBio™
QP7260-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_001031880.1 | |
S. aureus LDH-A / LDHA Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
31213039 | |
Research Use Only | |
LDHA | |
Recombinant Protein | |
E. coli | |
MNKFKGNKVV LIGNGAVGSS YAFSLVNQSI VDELVIIDLD TEKVRGDVMD LKHATPYSPT TVRVKAGEYS DCHDADLVVI CAGAAQKPGE TRLDLVSKNL KIFKSIVGEV MASKFDGIFL VATNPVDILA YATWKFSGLP KERVIGSGTI LDSARFRLLL SEAFDVAPRS VDAQIIGEHG DTELPVWSHA NIAGQPLKTL LEQRPEGKAQ IEQIFVQTRD AAYDIIQAKG ATYYGVAMGL ARITEAIFRN EDAVLTVSAL LEGEYDEEDV YIGVPAVINR NGIRNVVEIP LNDEEQSKFA HSAKTLKDIM AEAEELK | |
Greater than 90% as determined by SDS-PAGE. |