missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Peptide deformylase Protein
A DNA sequence encoding the Staphylococcus aureus Peptide deformylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6478-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_000957037.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
def | |
Recombinant Protein | |
E. coli | |
MLTMKDIIRD GHPTLRQKAA ELELPLTKEE KETLIAMREF LVNSQDEEIA KRYGLRSGVG LAAPQINISK RMIAVLIPDD GSGKSYDYML VNPKIVSHSV QEAYLPTGEG CLSVDDNVAG LVHRHNRITI KAKDIEGNDI QLRLKGYPAI VFQHEIDHLN GVMFYDHIDK NHPLQPHTDA VEV |
S. aureus Peptide deformylase Protein | |
1 mg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |