missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Staphylococcal secretory antigen ssaA1 Protein
A DNA sequence encoding the Staphylococcus aureus (strain COL) Staphylococcal secretory antigen ssaA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9791-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_000725225.1 | |
1 mg | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
S. aureus Staphylococcal secretory antigen ssaA1 Protein | |
Research Use Only | |
ssaA1 | |
Recombinant Protein | |
Yeast | |
AEQNNNGYNS NDAQSYSYTY TIDAQGNYHY TWTGNWNPSQ LTQNNTYYYN NYNTYSYNNA SYNNYYNHSY QYNNYTNNSQ TATNNYYTGG SGASYSTTSN NVHVTTTAAP SSNGRSISNG YASGSNLYTS GQCTYYVFDR VGGKIGSTWG NASNWANAAA SSGYTVNNTP KVGAIMQTTQ GYYGHVAYVE GVNSNGSVRV SEMNYGHGAG VVTSRTISAN QAGSYNFIH | |
Greater than 90% as determined by SDS-PAGE. |