missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Proliferating cell nuclear antigen Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Proliferating cell nuclear antigen, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6471-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009645.1 | |
S. cerevisiae Proliferating cell nuclear antigen Protein | |
Research Use Only | |
POL30 | |
Recombinant Protein | |
E. coli | |
MLEAKFEEAS LFKRIIDGFK DCVQLVNFQC KEDGIIAQAV DDSRVLLVSL EIGVEAFQEY RCDHPVTLGM DLTSLSKILR CGNNTDTLTL IADNTPDSII LLFEDTKKDR IAEYSLKLMD IDADFLKIEE LQYDSTLSLP SSEFSKIVRD LSQLSDSINI MITKETIKFV ADGDIGSGSV IIKPFVDMEH PETSIKLEMD QPVDLTFGAK YLLDIIKGSS LSDRVGIRLS SEAPALFQFD LKSGFLQFFL APKFNDEE | |
Greater than 90% as determined by SDS-PAGE. |
852385 | |
50 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |