missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL6 / Interleukin-6 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6225-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009392.1 | |
IL6 / Interleukin-6 Protein | |
10 μg | |
Not determined. | |
Full Length | |
Sheep | |
GST | |
Tris-based buffer, 50% glycerol |
443406 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
E. coli | |
GPLGEDFKND TTPSRLLLTT PEKTEALIKH IVDKISAIRK EICEKNDECE NSKETLAENK LKLPKMEEKD GCFQSGFNQA ICLIKTTAGL LEYQIYLDFL QNEFEGNQET VMELQSSIRT LIQILKEKIA GLITTPATHT DMLEKMQSSN EWVKNAKVII ILRSLENFLQ FSLRAIRMK |