missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Viral CKBP Protein

Produktkod. 18786567
Change view
Klicka för att se tillgängliga alternativ
Formulering:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Formulering
18786567 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
18785687 Lyophilized from a 0.2μm filtered solution in PBS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18786567 Leverantör R&D Systems™ Leverantörsnummer 8254CK025
 Begränsad artikel
Beräknad leverans: 04-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Viral CKBP Protein is derived from HEK293. The Recombinant Viral CKBP Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer P19063
Formulering Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Molekylvikt (g/mol) 27kDa
Namn CKBP
Beredningsmetod Reconstitute at 500μg/mL in sterile PBS.
Kvalitetskontrolltestning SDS-PAGE: 36-40kDa, reducing conditions
Kvantitet 25 μg
Källa Human embryonic kidney cell,HEK293-derived viral CKBP protein Pro18-Val258,with an N-terminal 6-His tag
Förvaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatorisk status RUO
Uppsägning His
Konjugera Unconjugated
Rekombinant Recombinant
Produktlinje R&D Systems
Renhet eller kvalitetsklass 90%, by SDS-PAGE with silver staining
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.